siRNA Details
| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175275.4 | NP_780484.2 | 5 | 925 | ||
| NM_177385.4 | NP_796359.1 | 5 | 925 | |||
| GenBank mRNA | AK031604.1 | - | 5 | 831 | ||
| AK052558.1 | - | 5 | 845 | |||
| AK165111.1 | - | 4 | 398 | |||
| BC125622.1 | - | 5 | 800 | |||
| BC125624.1 | - | 5 | 800 | |||

Cntln
Genomic Map