siRNA Details
| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037906.2 | NP_001032995.1 | 19 | 2312 | ||
| GenBank mRNA | AK046127.1 | - | 1641 | |||
| AY622226.1 | - | 19 | 2312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nell1| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037906.2 | NP_001032995.1 | 19 | 2312 | ||
| GenBank mRNA | AK046127.1 | - | 1641 | |||
| AY622226.1 | - | 19 | 2312 | |||