siRNA Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045528.1 | NP_001038993.1 | 19 | 2017 | ||
| GenBank mRNA | AK049459.1 | - | 19 | 1962 | ||
| AK049509.1 | - | 19 | 2017 | |||
| AK085087.1 | - | 19 | 2008 | |||
| AK122343.1 | - | 12 | 1256 | |||
| AK161705.1 | - | 19 | 1947 | |||
| BC034098.1 | - | 9 | 954 | |||
| BC060651.1 | - | 19 | 1931 | |||

Fkbp15
Genomic Map