siRNA Details
| Gene Symbol: Tmem241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289666.1 | NP_001276595.1 | 5 | 382 | ||
| NM_178801.5 | NP_848916.2 | 5 | 382 | |||
| GenBank mRNA | AK031517.1 | - | 5 | 376 | ||
| AK038419.1 | - | 5 | 360 | |||
| AK038472.1 | - | 5 | 327 | |||
| AK051727.1 | - | 6 | 533 | |||
| AK144963.1 | - | 5 | 384 | |||
| BC094382.1 | - | 5 | 298 | 30605060 | ||
| BC137621.1 | - | 5 | 312 | 8860640 | ||

Tmem241
Genomic Map