siRNA Details
| Gene Symbol: Trim65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178802.4 | NP_848917.2 | 6 | 1297 | ||
| GenBank mRNA | AK028857.1 | - | 6 | 1186 | ||
| AK047439.1 | - | 6 | 1297 | |||
| AK050080.1 | - | 6 | 1209 | |||
| AK079982.1 | - | 5 | 1379 | |||
| BC118954.1 | - | 6 | 1052 | |||
| BC118955.1 | - | 6 | 1052 | |||

Trim65
Genomic Map