siRNA Details
| Gene Symbol: Mia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177389.3 | NP_796363.2 | 8, 9 | 3740 | ||
| GenBank mRNA | AK046506.1 | - | 3, 4 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mia3| Gene Symbol: Mia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177389.3 | NP_796363.2 | 8, 9 | 3740 | ||
| GenBank mRNA | AK046506.1 | - | 3, 4 | 476 | ||