siRNA Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177391.4 | NP_796365.1 | 2 | 579 | ||
| GenBank mRNA | AK083325.1 | - | 2 | 554 | ||
| AK090060.1 | - | 2 | 674 | |||
| BC116256.1 | - | 1 | 347 | 40093730 | ||
| BC116257.1 | - | 1 | 344 | 40093731 | ||

Fam109b
Genomic Map