siRNA Details
| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177392.3 | NP_796366.1 | 3, 4 | 218 | ||
| GenBank mRNA | AK043094.1 | - | 3, 4 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem220| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177392.3 | NP_796366.1 | 3, 4 | 218 | ||
| GenBank mRNA | AK043094.1 | - | 3, 4 | 219 | ||