siRNA Details
| Gene Symbol: Map3k9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174107.1 | NP_001167578.1 | 2, 3 | 1098 | ||
| NM_177395.5 | NP_796369.2 | 2, 3 | 1098 | |||
| GenBank mRNA | AK053843.1 | - | 2, 3 | 939 | ||
| AK155677.1 | - | 2, 3 | 1104 | |||

Map3k9
Genomic Map