siRNA Details
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 8 | 1156 | ||
GenBank mRNA | AK085308.1 | - | 8 | 1146 | ||
BC043305.1 | - | 8 | 1085 | 5040944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 8 | 1156 | ||
GenBank mRNA | AK085308.1 | - | 8 | 1146 | ||
BC043305.1 | - | 8 | 1085 | 5040944 |