siRNA Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178086.3 | NP_835187.2 | 6 | 1092 | ||
| GenBank mRNA | AK033736.1 | - | 6 | 1090 | ||
| AK090338.1 | - | 6 | 1086 | |||
| AK161502.1 | - | 6 | 1090 | |||
| AY660882.1 | - | 6 | 1023 | |||
| BC026400.1 | - | 5 | 567 | |||
| BC026629.1 | - | 4 | 634 | |||
| BC046985.1 | - | 6 | 1048 | |||
| BC111912.1 | - | 7 | 913 | |||
| BC128080.1 | - | 6 | 1073 | 40104374 | ||
| BC128081.1 | - | 6 | 1073 | 40104375 | ||

Fa2h
Genomic Map