siRNA Details
| Gene Symbol: Pramel6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178249.2 | NP_839980.1 | 3 | 1063 | ||
| GenBank mRNA | AF490342.1 | - | 3 | 1063 | ||
| AK139830.1 | - | 4 | 1297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramel6| Gene Symbol: Pramel6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178249.2 | NP_839980.1 | 3 | 1063 | ||
| GenBank mRNA | AF490342.1 | - | 3 | 1063 | ||
| AK139830.1 | - | 4 | 1297 | |||