siRNA Details
| Gene Symbol: Pramel7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178250.2 | NP_839981.1 | 1 | 88 | ||
| GenBank mRNA | AF490343.1 | - | 1 | 88 | ||
| BC052825.1 | - | 1 | 97 | 30030731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramel7| Gene Symbol: Pramel7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178250.2 | NP_839981.1 | 1 | 88 | ||
| GenBank mRNA | AF490343.1 | - | 1 | 88 | ||
| BC052825.1 | - | 1 | 97 | 30030731 | ||