siRNA Details
| Gene Symbol: Plekhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183034.1 | NP_898855.1 | 5 | 1204 | ||
| GenBank mRNA | AK154703.1 | - | 4 | 1133 | ||
| AK155357.1 | - | 5 | 1228 | |||
| BC053079.1 | - | 5 | 1204 | 5702529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhm1| Gene Symbol: Plekhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183034.1 | NP_898855.1 | 5 | 1204 | ||
| GenBank mRNA | AK154703.1 | - | 4 | 1133 | ||
| AK155357.1 | - | 5 | 1228 | |||
| BC053079.1 | - | 5 | 1204 | 5702529 | ||