siRNA Details
| Gene Symbol: Tas2r123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207025.1 | NP_996908.1 | 1 | 455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r123| Gene Symbol: Tas2r123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207025.1 | NP_996908.1 | 1 | 455 | ||