siRNA Details
| Gene Symbol: Eras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181548.2 | NP_853526.1 | 2 | 358 | ||
| GenBank mRNA | AB093573.1 | - | 2 | 358 | ||
| AK141212.1 | - | 2 | 252 | |||
| BC119277.1 | - | 2 | 274 | 8734026 | ||
| BC119281.1 | - | 2 | 274 | 8734030 | ||

Eras
Genomic Map