siRNA Details
| Gene Symbol: Zfp703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101502.1 | NP_001094972.1 | 2 | 757 | ||
| NM_001110508.1 | NP_001103978.1 | 3 | 891 | |||
| GenBank mRNA | BE981624.1 | - | 1 | 315 | ||
| BF464968.1 | - | 1 | 316 | |||
| JF713461.1 | - | 2 | 739 | |||

Zfp703
Genomic Map