siRNA Details
| Gene Symbol: Rtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184109.1 | NP_908998.1 | 1 | 1973 | ||
| GenBank mRNA | EU434913.1 | - | 2095 | |||
| EU434918.1 | - | 2120 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtl1| Gene Symbol: Rtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184109.1 | NP_908998.1 | 1 | 1973 | ||
| GenBank mRNA | EU434913.1 | - | 2095 | |||
| EU434918.1 | - | 2120 | ||||