siRNA Details
| Gene Symbol: Gpr141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181754.4 | NP_861419.3 | 5 | 981 | ||
| GenBank mRNA | AK154688.1 | - | 5 | 982 | ||
| AY288427.1 | - | 1 | 470 | |||
| BC125585.1 | - | 1 | 554 | 40130100 | ||
| BC125587.1 | - | 1 | 554 | 40130102 | ||
More Information
Set Membership: |
|

Gpr141
Genomic Map