siRNA Details
| Gene Symbol: Tmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181820.2 | NP_861541.2 | 14 | 2024 | ||
| GenBank mRNA | AK050105.1 | - | 13 | 1919 | ||
| AY236493.1 | - | 14 | 1987 | |||
| AY263162.1 | - | 14 | 1995 | |||
| BC022758.1 | - | 6 | 714 | |||
| BC040466.1 | - | 14 | 1936 | |||
| CN719678.1 | - | 3 | 218 | |||

Tmc4
Genomic Map