siRNA Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY241873.1 | - | 574 | |||
| AY241874.1 | - | 1008 | ||||
| AY369078.2 | - | 80 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm1| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY241873.1 | - | 574 | |||
| AY241874.1 | - | 1008 | ||||
| AY369078.2 | - | 80 | ||||