siRNA Details
| Gene Symbol: Defb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183035.1 | NP_898856.1 | 2 | 92 | ||
| GenBank mRNA | AJ578469.1 | - | 2 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb34| Gene Symbol: Defb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183035.1 | NP_898856.1 | 2 | 92 | ||
| GenBank mRNA | AJ578469.1 | - | 2 | 92 | ||