siRNA Details
| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138628.3 | NP_619534.2 | 10 | 1518 | ||
| GenBank mRNA | AF422244.1 | - | 10 | 1455 | ||
| AF422245.1 | - | 10 | 1455 | |||
| AK084639.1 | - | 10 | 1519 | |||
| AK085904.1 | - | 10 | 1414 | |||
| AK137442.1 | - | 10 | 1433 | |||
| BC141023.1 | - | 10 | 1431 | 9055635 | ||

Txlnb
Genomic Map