siRNA Details
| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138628.3 | NP_619534.2 | 9 | 1452 | ||
| GenBank mRNA | AF422244.1 | - | 9 | 1389 | ||
| AF422245.1 | - | 9 | 1389 | |||
| AK084639.1 | - | 9 | 1453 | |||
| AK085904.1 | - | 9 | 1348 | |||
| AK137442.1 | - | 9 | 1367 | |||
| BC141023.1 | - | 9 | 1365 | 9055635 | ||

Txlnb
Genomic Map