siRNA Details
| Gene Symbol: Bpifb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194357.1 | NP_919338.2 | 2 | 228 | ||
| GenBank mRNA | BC048084.1 | - | 2 | 228 | 6312261 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifb3| Gene Symbol: Bpifb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194357.1 | NP_919338.2 | 2 | 228 | ||
| GenBank mRNA | BC048084.1 | - | 2 | 228 | 6312261 | |