siRNA Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198119.2 | NP_932787.1 | 6 | 1599 | ||
| GenBank mRNA | AK048678.1 | - | 6 | 1592 | ||
| AK078176.1 | - | 6 | 1600 | |||
| BC116884.1 | - | 4 | 1391 | 40126203 | ||
| BC116886.1 | - | 4 | 1391 | 40126205 | ||

Lrrc24
Genomic Map