siRNA Details
| Gene Symbol: Cfap54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC046559.1 | - | 5 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap54| Gene Symbol: Cfap54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC046559.1 | - | 5 | 664 | ||