siRNA Details
| Gene Symbol: Lemd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081193.2 | NP_001074662.2 | 6 | 1930 | ||
| GenBank mRNA | AK140538.1 | - | 6 | 413 | ||
| BC082610.1 | - | 7 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lemd3| Gene Symbol: Lemd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081193.2 | NP_001074662.2 | 6 | 1930 | ||
| GenBank mRNA | AK140538.1 | - | 6 | 413 | ||
| BC082610.1 | - | 7 | 694 | |||