siRNA Details
| Gene Symbol: Slc47a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033542.2 | NP_001028714.1 | 16 | 1527 | ||
| GenBank mRNA | AK133432.1 | - | 16 | 1528 | ||
| BC147619.1 | - | 16 | 1475 | 9056609 | ||
| BC157913.1 | - | 16 | 1475 | 9088130 | ||

Slc47a2
Genomic Map