siRNA Details
| Gene Symbol: Tmem102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033433.4 | NP_001028605.2 | 2 | 927 | ||
| GenBank mRNA | AK143262.1 | - | 2 | 927 | ||
| BC144871.1 | - | 1 | 977 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem102| Gene Symbol: Tmem102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033433.4 | NP_001028605.2 | 2 | 927 | ||
| GenBank mRNA | AK143262.1 | - | 2 | 927 | ||
| BC144871.1 | - | 1 | 977 | |||