siRNA Details
| Gene Symbol: Scarf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004157.2 | NP_001004157.2 | 11 | 2199 | ||
| GenBank mRNA | AK133477.1 | - | 11 | 2182 | ||
| AK220215.1 | - | 11 | 2183 | |||
| BC057600.1 | - | 10 | 2106 | |||
| BC080288.1 | - | 9 | 1930 | 6411836 | ||

Scarf1
Genomic Map