siRNA Details
| Gene Symbol: Ccdc177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008423.2 | NP_001008423.2 | 2 | 2906 | ||
| GenBank mRNA | AK147473.1 | - | 2733 | |||
| BC067044.1 | - | 2 | 2878 | 30531780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc177| Gene Symbol: Ccdc177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008423.2 | NP_001008423.2 | 2 | 2906 | ||
| GenBank mRNA | AK147473.1 | - | 2733 | |||
| BC067044.1 | - | 2 | 2878 | 30531780 | ||