siRNA Details
| Gene Symbol: Igh-VJ558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF296427.1 | - | 1 | 166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igh-VJ558| Gene Symbol: Igh-VJ558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF296427.1 | - | 1 | 166 | ||