siRNA Details
| Gene Symbol: Gm5141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256065.1 | NP_001242994.1 | 4 | 1562 | ||
| GenBank mRNA | BC064714.1 | - | 4 | 1562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5141| Gene Symbol: Gm5141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256065.1 | NP_001242994.1 | 4 | 1562 | ||
| GenBank mRNA | BC064714.1 | - | 4 | 1562 | ||