siRNA Details
| Gene Symbol: Gm906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033438.2 | NP_001028610.1 | 4 | 1521 | ||
| GenBank mRNA | AK133043.1 | - | 4 | 1522 | ||
| BC147515.1 | - | 4 | 1504 | 9056507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm906| Gene Symbol: Gm906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033438.2 | NP_001028610.1 | 4 | 1521 | ||
| GenBank mRNA | AK133043.1 | - | 4 | 1522 | ||
| BC147515.1 | - | 4 | 1504 | 9056507 | ||