siRNA Details
| Gene Symbol: Gm906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033438.2 | NP_001028610.1 | 4 | 1635 | ||
| GenBank mRNA | AK133043.1 | - | 4 | 1636 | ||
| BC147515.1 | - | 4 | 1618 | 9056507 | ||
| Gene Symbol: Gm8765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244649.1 | NP_001231578.1 | 4 | 1618 | ||
| GenBank mRNA | BC172081.1 | - | 4 | 1618 | 9054367 | |

Gm906
Genomic Map