siRNA Details
| Gene Symbol: Zfp395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199029.2 | NP_950194.2 | 10 | 1758 | ||
| GenBank mRNA | AK155538.1 | - | 10 | 1759 | ||
| BC053701.1 | - | 825 | 30054630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp395| Gene Symbol: Zfp395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199029.2 | NP_950194.2 | 10 | 1758 | ||
| GenBank mRNA | AK155538.1 | - | 10 | 1759 | ||
| BC053701.1 | - | 825 | 30054630 | |||