siRNA Details
| Gene Symbol: Siah3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128093.1 | NP_001121565.1 | 2 | 617 | ||
| GenBank mRNA | BC147762.1 | - | 2 | 617 | 9087487 | |
| BC147764.1 | - | 2 | 617 | 9087489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siah3| Gene Symbol: Siah3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128093.1 | NP_001121565.1 | 2 | 617 | ||
| GenBank mRNA | BC147762.1 | - | 2 | 617 | 9087487 | |
| BC147764.1 | - | 2 | 617 | 9087489 | ||