siRNA Details
| Gene Symbol: Kmt2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033276.3 | NP_001028448.3 | 46, 47 | 14775 | ||
| GenBank mRNA | BC058659.1 | - | 7, 8 | 1534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2d| Gene Symbol: Kmt2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033276.3 | NP_001028448.3 | 46, 47 | 14775 | ||
| GenBank mRNA | BC058659.1 | - | 7, 8 | 1534 | ||