siRNA Details
| Gene Symbol: Parl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005767.4 | NP_001005767.1 | 10 | 1128 | ||
| GenBank mRNA | AK150980.1 | - | 10 | 1134 | ||
| AK151652.1 | - | 10 | 1127 | |||
| AK207495.1 | - | 1 | 60 | |||
| AK218894.1 | - | 1 | 60 | |||
| BC083153.1 | - | 10 | 1128 | 30728006 | ||

Parl
Genomic Map