siRNA Details
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034891.3 | NP_001030063.1 | 4 | 528 | ||
| NM_001039552.2 | NP_001034641.2 | 1461 | ||||
| GenBank mRNA | AK019108.1 | - | 5 | 616 | ||
| AK160176.1 | - | 4 | 529 | |||
| BC150877.1 | - | 1522 | 9087788 | |||

Ermard
Genomic Map