siRNA Details
| Gene Symbol: Cdkl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033443.4 | NP_001028615.1 | 6 | 800 | ||
| GenBank mRNA | AK157995.1 | - | 6 | 801 | ||
| BC138986.1 | - | 5 | 698 | 8862008 | ||
| BC138988.1 | - | 5 | 698 | 8862010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkl4| Gene Symbol: Cdkl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033443.4 | NP_001028615.1 | 6 | 800 | ||
| GenBank mRNA | AK157995.1 | - | 6 | 801 | ||
| BC138986.1 | - | 5 | 698 | 8862008 | ||
| BC138988.1 | - | 5 | 698 | 8862010 | ||