siRNA Details
| Gene Symbol: Capn13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033444.2 | NP_001028616.1 | 10 | 1130 | ||
| GenBank mRNA | AK136569.1 | - | 10 | 1131 | ||
| BC147510.1 | - | 10 | 1047 | 9056477 | ||
| BC147512.1 | - | 10 | 1047 | 9056488 | ||

Capn13
Genomic Map