siRNA Details
| Gene Symbol: Ccdc68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201362.2 | NP_958750.1 | 5 | 467 | ||
| GenBank mRNA | AK078964.1 | - | 5 | 469 | ||
| BC046343.1 | - | 5 | 447 | 6487853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc68| Gene Symbol: Ccdc68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201362.2 | NP_958750.1 | 5 | 467 | ||
| GenBank mRNA | AK078964.1 | - | 5 | 469 | ||
| BC046343.1 | - | 5 | 447 | 6487853 | ||