siRNA Details
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201362.2 | NP_958750.1 | 6 | 656 | ||
GenBank mRNA | AK078964.1 | - | 6 | 658 | ||
BC046343.1 | - | 6 | 636 | 6487853 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201362.2 | NP_958750.1 | 6 | 656 | ||
GenBank mRNA | AK078964.1 | - | 6 | 658 | ||
BC046343.1 | - | 6 | 636 | 6487853 |