siRNA Details
| Gene Symbol: Tmem151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001885.1 | NP_001001885.1 | 2 | 467 | ||
| GenBank mRNA | AK139456.1 | - | 2 | 522 | ||
| AK144531.1 | - | 2 | 478 | |||
| AK155690.1 | - | 2 | 484 | |||
| AK163651.1 | - | 2 | 484 | |||
| BC072634.1 | - | 2 | 467 | 6850125 | ||

Tmem151a
Genomic Map