siRNA Details
| Gene Symbol: Ap5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033448.2 | NP_001028620.1 | 2 | 2223 | ||
| GenBank mRNA | AK144121.1 | - | 2 | 2226 | ||
| AK171710.1 | - | 3 | 1919 | |||
| BC147482.1 | - | 2 | 2146 | 9056228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap5b1| Gene Symbol: Ap5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033448.2 | NP_001028620.1 | 2 | 2223 | ||
| GenBank mRNA | AK144121.1 | - | 2 | 2226 | ||
| AK171710.1 | - | 3 | 1919 | |||
| BC147482.1 | - | 2 | 2146 | 9056228 | ||