siRNA Details
| Gene Symbol: Ap5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033448.2 | NP_001028620.1 | 1, 2 | 533 | ||
| GenBank mRNA | AK144121.1 | - | 1, 2 | 536 | ||
| AK171710.1 | - | 2, 3 | 229 | |||
| BC147482.1 | - | 1, 2 | 456 | 9056228 | ||

Ap5b1
Genomic Map