siRNA Details
| Gene Symbol: Fam189a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114174.1 | NP_001107646.1 | 10 | 1405 | ||
| GenBank mRNA | BC099492.1 | - | 5 | 529 | ||
| DV649334.1 | - | 5 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189a2| Gene Symbol: Fam189a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114174.1 | NP_001107646.1 | 10 | 1405 | ||
| GenBank mRNA | BC099492.1 | - | 5 | 529 | ||
| DV649334.1 | - | 5 | 500 | |||