siRNA Details
| Gene Symbol: 4430402I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198651.2 | NP_941053.2 | 10, 11 | 1330 | ||
| GenBank mRNA | AK031997.1 | - | 10, 11 | 1330 | ||
| BC147044.1 | - | 9, 10 | 910 | 9056033 | ||
| BC147045.1 | - | 9, 10 | 910 | 9056045 | ||
| BC171985.1 | - | 8, 9 | 859 | 9054271 | ||
| BC171986.1 | - | 8, 9 | 868 | 9054272 | ||

4430402I18Rik
Genomic Map